Restek™ Mobile Phase Degasser
Uses tubing composed of 200 to 300 times more gas permeable amorphous fluoropolymeter than PTFE
Manufacturer: Restek™ 25189
 View more versions of this product
Catalog No. 06-712-745
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Mobile Phase Degasser
    Includes 4 channels
    Pressure 0.24psi (loss)
    Voltage 110V
    Volume (Metric) 500mL
  • Description & Specifications
Related Products
Product Content Feedback