Learn More
Restek Mobile Phase Inlet Filters and Helium Spargers
Protect HPLC system from small particles
$47.50 - $48.50
Catalog Number | Mfr. No. | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Porosity | Price | Quantity & Availability | |||||
06-712-854
|
Restek
25311 |
2 µm |
Each for $48.50
|
|
06-712-855
|
Restek
25312 |
10 µm |
Each for $47.50
|
|
06-712-856
|
Restek
25313 |
20 µm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Type 316 stainless steel and PEEK
- Compatible with most solvents
- When used as helium sparger, offer an inexpensive way to prepare and maintain mobile phases, free of dissolved gas
Specifications
Restek Mobile Phase Inlet Filter |