Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ MXT™-1 Capillary Columns, 15m Total Length Available on GSA/VA Contract for Federal Government customers only.

Crossbond 100% dimethyl polysiloxane

Manufacturer:  Restek™ 70182104

 View more versions of this product

Catalog No. 06-713-505


  Check Availability
Add to cart

Specifications

Description

Applications include solvent impurities, PCB congeners
Description & Specifications

Specifications

Solvent impurities, PCB congeners, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
445°C
3μm
3μm
Stainless Steel
100% Dimethyl Polysiloxane
Non polar
0.53mm
Capillary Column
G1, G2, G38
-60 to 285/360°C
15m
Crossbond™
Mxt™-1
Siltek™
1 Ea.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit