Learn More
Restek™ MXT-1 Silcosteel Capillary Columns - 6m Length Available on GSA/VA Contract for Federal Government customers only.
Siltek™-treated stainless steel
Manufacturer:  Restek™ 70101
 View more versions of this product
Catalog No. 06-713-286
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvent impurities, PCB congeners, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
G1, G2, G38
-60 to 430°C
6m
Crossbond™
Mxt™-1
Siltek™
445°C
0.15μm
0.15μm
Stainless Steel
100% Dimethyl Polysiloxane
Non polar
0.53mm