Learn More
Restek™ MXT-200 Silcosteel Capillary Columns, 40m and Longer Available on GSA/VA Contract for Federal Government customers only.
Mid-polarity phase, Crossbond™ trifluoropropylmethyl polysiloxane
Manufacturer:  Restek™ 75058283
 View more versions of this product
Catalog No. 06-714-138
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvents, Freon™ Fluorocarbons, Alcohols, Ketones, Silanes, Glycols analysis
G6
-20 to 290/360°C
Solute stationary phase interaction
Stainless Steel
Trifluoropropylmethyl Polysiloxane
Mid polar
0.53mm
445°C
1μm
1μm
60m
Crossbond™
Mxt™-200
Siltek™
3.5 in. Coil