Learn More
Restek™ MXT™-502.2 Silcosteel™ Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Siltek™-treated stainless steel
Manufacturer:  Restek™ 70908273
 View more versions of this product
Catalog No. 06-713-925
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatile organic pollutants, Trihalomethanes, Light Hydrocarbons, Aromatics analysis
3μm
3μm
Stainless Steel
Proprietary Diphenyl Dimethyl Polysiloxane
Siltek™
3.5 in. Coil
445°C
-20 to 270/320°C
30m
Crossbond™
Mxt™-502.2
0.53mm