Learn More
Restek™ MXT™-WAX Capillary Columns: 20m Available on GSA/VA Contract for Federal Government customers only.
Crossbond Carbowax polyethylene glycol.
Manufacturer:  Restek™ 70655112
 View more versions of this product
Catalog No. 06-713-858
  Check Availability
Add to cart
Specifications
Description
Applications include FAMEs, flavor compounds, essential oils.
Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, Amines, solvents, Xylene Isomers analysis
445°C
1μm
1μm
Stainless Steel
Carbowax™ Polyethylene Glycol
Polar
0.53mm
1 Ea.
Capillary Column
G14, G15, G16, G20, G39
40 to 240/250°C
30m
Crossbond™
Mxt™-Wax
Siltek™
Not Applicable