Learn More
Restek™ MXT™-WAX Capillary Columns, 30m Available on GSA/VA Contract for Federal Government customers only.
General-purpose columns for FAMEs, flavor compounds, essential oils, amines, solvents, xylene isomers, and U.S. EPA Method 603 (acrolein/acrylonitrile).
Manufacturer:  Restek™ 70655283
 View more versions of this product
Catalog No. 06-713-864
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, Amines, solvents, Xylene Isomers analysis
G14, G15, G16, G20, G39
40 to 240/250°C
30m
Crossbond™
Mxt™-Wax
Siltek™
3.5 in. Coil
445°C
1μm
1μm
Stainless Steel
Carbowax™ Polyethylene Glycol
Polar
0.53mm