Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ MXT-WAX Silcosteel Capillary Columns: 30m Available on GSA/VA Contract for Federal Government customers only.

General-purpose columns for FAMEs, flavor compounds, essential oils, amines, solvents, xylene isomers, and U.S. EPA Method 603 (acrolein/acrylonitrile).

Manufacturer:  Restek™ 70625273

 View more versions of this product

Catalog No. 06-713-784


  Check Availability
Add to cart

Specifications

Description

  • Polar phase
  • Crossbond™ Carbowax polyethylene glycol phase provides oxidation resistance
  • Siltek treated
  • Equivalent to USP G14, G15, G16, G20, and G39 phases
  • Standard coil diameter: 4.5 in.
  • Order Info

    Custom lengths and film thicknesses available.

    Description & Specifications

    Specifications

    FAMEs, flavor compounds, essential oils, Amines, solvents, Xylene Isomers analysis
    G14, G15, G16, G20, G39
    40 to 250/260°C
    30m
    Crossbond™
    Mxt™-Wax
    Siltek™
    3.5 in. Coil
    445°C
    0.25μm
    0.25μm
    Stainless Steel
    Carbowax™ Polyethylene Glycol
    Polar
    0.53mm
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit