Learn More
Restek™ Normal Phase Resprep SPE Cartridges
Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices.
Manufacturer:  Restek™ 26085
 View more versions of this product
Catalog No. 06-713-146
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Hydrophilic (Polar)
Normal
Florisil™
SPE cartridge
1000mg/6mL
HDPE, Polyethylene Frits
Used to extract hydrophilic analytes from non-Polar matrices, such as organic solvents (e.g., Polar contaminants from sample extracts)
Florisil™
6mL, 1,000mg