Learn More
Restek™ Normal Phase Resprep SPE Cartridges
Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices.
Manufacturer:  Restek™ 24032
 View more versions of this product
Catalog No. 06-718-812
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Hydrophilic (Polar)
Normal
Florisil™
SPE cartridge
500mg/3mL
HDPE, Polyethylene Frits
Used to extract hydrophilic analytes from non-Polar matrices, such as organic solvents (e.g., Polar contaminants from sample extracts)
Florisil™ PTFE frits
3mL, 500mg