Restek™ Normal Phase Resprep SPE Cartridges
Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices.
Manufacturer: Restek™ 24034
 View more versions of this product
Catalog No. 06-718-814
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Adsorbent Type Hydrophilic (Polar)
    Cartridge Material HDPE, Polyethylene Frits
    Column Phase Normal
    For Use With (Application) Used to extract hydrophilic analytes from non-Polar matrices, such as organic solvents (e.g., Polar contaminants from sample extracts)
    Material Florisil™
    Packing Material Florisil™ PTFE frits
    Tube Size 6mL, 1,000mg
    Volume (Metric) 1000mg/6mL
  • Description & Specifications
Related Products
Product Content Feedback