Learn More
Restek™ Oil-Free Vacuum/Pressure Station for Resprep Systems
For 12-Port Resprep™ SPE Systems.
Manufacturer:  Restek™ 24002
 View more versions of this product
Catalog No. 06-712-140
  Check Availability
Add to cart
Specifications
Description
  • Allows switching from pressure to vacuum in a matter of seconds
  • Quiet, oil-free unit will not contaminate the extraction system or samples
  • Provides 20 in. Hg (68 kPa) vacuum or 18psi (124 kPa) pressure
Order Info
Not recommended for use with 24-port manifold.
Description & Specifications
Specifications
115V 60Hz
12 Port Resprep™ SPE system
18psi
115V
To allow switching from pressure to vacuum
60Hz
Oil free vacuum/pressure station