Learn More
Restek™ One-Piece Fused Silica Adaptor Ferrule
Fused silica adaptor ferrules made from Valcon polyimide or virgin polyimide.
Supplier: Restek™ 20142
Description
Fused silica adaptor ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C. The determining factor for selecting adaptor ferrule size is the fused silica tubing OD.Specifications
Valcon Polyimide, Fused Silica | |
0.003 to 0.031 in. | |
350°C | |
0.025 to 0.04 cm | |
0.158 cm |
662°F | |
0.0625 in. | |
0.009 to 0.015 in. | |
0.01 to 0.025 cm | |
Valco Adaptor Ferrule |