Learn More
Restek™ One-Piece Fused Silica Adaptor Ferrule
Fused silica adaptor ferrules made from Valcon polyimide or virgin polyimide.
Manufacturer:  Restek™ 20137
 View more versions of this product
Catalog No. 06-717-595
  Check Availability
Add to cart
Specifications
Description
Fused silica adaptor ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C. The determining factor for selecting adaptor ferrule size is the fused silica tubing OD.
Description & Specifications
Specifications
0.031 in.
0.025cm
0.0312 in.
662°F
Valco™ adaptor ferrule
<0.009 to 0.015 in.
<0.025 to 0.04cm
0.079cm
350°C
Valcon Polyimide, Fused Silica