Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Opti-Cap Bottle Top Kits

Fits all standard GL-45 bottles.

Manufacturer:  Restek™ 25302

 View more versions of this product

Catalog No. 06-712-845


  Check Availability
Add to cart

Includes: 3m of tubing, 2µm and 10µm sparging filters, 1L bottle

Specifications

Description

  • Economical way to helium-sparge and deliver LC mobile phases
  • Have two 0.125 in. holes and one 0.0625 in. hole for tubing
  • All three openings are designed for threaded PEEK plugs
Part No. 25301: Includes 3m of tubing, 2µm and 10µm sparging filters
Part No. 25302: Includes 3m of tubing, 2µm and 10µm sparging filters, 1L bottle
Part No. 25303: Includes 3m of tubing, 2µm and 10µm sparging filters, 2L bottle
Description & Specifications

Specifications

Opti-Cap™ Kit
3m of tubing, 2μm and 10μm sparging filters, 1L bottle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit