Learn More
Restek™ Pinnacle™ DB Biphenyl HPLC Prep Columns, 5um Particle Size
An excellent choice for the analysis of steroids, tetracyclines, drug metabolites, and other compounds that contain some degree of unsaturation.
Manufacturer:  Restek™ 9409519
 View more versions of this product
Catalog No. 06-722-307
  Check Availability
Add to cart
Specifications
Description
Pinnacle DB Biphenyl is a unique reversed phase material that displays both increased retention and selectivity for aromatic and/or unsaturated compounds when compared to conventional alkyl and phenyl phases. Highly base-deactivated spherical silica. Endcapped.
Description & Specifications
Specifications
8%
30mm
5μm
Pi-Pi
100mm
Silica
5μm
140Å
150m2/g
HPLC Column
Reverse phase HPLC
Phenyl groups
Yes
For Aromatic and unsaturated compounds
Yes
Biphenyl
Spherical
2.5 to 8
1 Ea.
80°C
L11
Pinnacle™ DB Biphenyl