Learn More
Restek™ Pinnacle DB Columns with C8 Column Packing
Base-deactivated spherical silica optimized for UHPLC stability.
Manufacturer:  Restek™ 9413562700
 View more versions of this product
Catalog No. 06-728-400
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6%
2.1mm
5μm
Analysis of basic compounds or bases mixed with Acids/neutrals
150mm
Stainless Steel
Silica
5μm
140Å
80°C
L7
Pinnacle™ DB C8
Pinnacle™ DB C8
Yes
0.2mL/min.
Trident™ inlet fitting
Monomeric C8
4000psi
Spherical
2.5 to 8
5g
HPLC Column
UHPLC