Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Pinnacle™ DB Cyano (CN) Mixed Mode HPLC Column with Trident Inlet Fitting, 5 μm
Carbon Load: 4%; Pore Size: 140 Å; Column Phase: Normal and Reversed Phase
Supplier: Restek™ 9416525700
Description
Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase and HILIC modes. Ideal for analysis of protonated bases.
- Particle Size: 5μm, spherical
- Pore Size: 140 angstrom
- Carbon Load: 4%
- End-Cap: yes
- pH Range: 2.5 to 8
- Temperature Limit: 80°C
- USP Phase Code: L10
- Phase Category: Cyano
- Ligand Type: Cyanopropylsilane
Specifications
4.6 mm | |
1 mL/min | |
Electrostatic/Dipole | |
200 mm | |
Spherical | |
2.5 to 8 | |
5 g | |
L10 | |
Silica | |
Mixed Mode | |
Normal and Reversed Phase | |
Column |
5 μm | |
Analysis of a Wide Range of Compounds, From Acidic Through Slightly Basic | |
Trident™ inlet fitting | |
Cyanopropylsilane | |
5 μm | |
140 Å | |
80°C | |
Cyano (CN) | |
4% | |
Yes | |
Stainless Steel | |
UHPLC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction