Learn More
Restek™ Pinnacle™ DB PFP Propyl HPLC Columns
Highly base-deactivated spherical silica
Manufacturer:  Restek™ 9419212
 View more versions of this product
Catalog No. 06-722-052
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6%
2.1mm
LC/MS
Spherical
2.5 to 8
80°C
L43
PFP Propyl
Yes
100mm
1.9μm
140Å
HPLC Column
PFP Propyl