Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Pinnacle™ II C18 HPLC Columns, 3µm Particle Size

Proprietary silica undergoes stringent testing onsite during manufacture, stationary phase bonding, and column packing

Manufacturer:  Restek™ 9214315700

 View more versions of this product

Catalog No. 06-717-209


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

13%
4.6mm
Acidic to neutral hydrophobic compounds
Spherical
2.5 to 8
80°C
L1
Pinnacle™ II C18
Yes
100mm
3μm
110Å
HPLC Column
HPLC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit