Learn More
Restek™ Pinnacle™ II C18 HPLC Columns, 5um Particle Size
SDP
Proprietary silica undergoes stringent testing onsite during manufacture, stationary phase bonding, and column packing
Supplier:  Restek™ 9214564
 View more versions of this product
Catalog No. 06-717-273
Add to Cart
Description
Description
  • Exhibit consistent quality and reproducibility
  • Narrower columns with faster flowrates decrease analysis times
  • Fully endcapped
  • Ideal for general-purpose, non-basic compound analysis
Specifications
Specifications
4 mm
Wide Range of Acidic to Neutral Hydrophobic Compounds
150 mm
Spherical
2.5 to 8
5 g
L1
Silica
Reversed Phase
Reversed Phase
5 μm
Trident™ inlet fitting
4000 psi (275 bar)
5 μm
110 Å
80°C
C18
13%
Yes
Column
Product Suggestions
Product Suggestions