Learn More
Restek™ Pinnacle™ II C18 HPLC Columns, 5um Particle Size
Proprietary silica undergoes stringent testing onsite during manufacture, stationary phase bonding, and column packing
Manufacturer:  Restek™ 9214512
 View more versions of this product
Catalog No. 06-717-240
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
13%
2.1mm
5μm
Wide range of Acidic to neutral hydrophobic compounds
100mm
Spherical
2.5 to 8
5g
HPLC Column
HPLC
Pinnacle™ II C18
Yes
0.2mL/min.
Trident™ inlet fitting
4000psi
5μm
110Å
80°C
L1
Pinnacle™ II C18