Learn More
Restek™ Pinnacle II™ Guard Cartridges
Manufacturer:  Restek™ 921450222
 View more versions of this product
Catalog No. 06-719-215
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
General purpose HPLC column
L1
Spherical
110Å
20mm
2.1mm
80°C
13%
Yes
3 or 5μm
2.5 to 8
3 Pk.
C18