Learn More
Restek™ Pinnacle™ II Silica HPLC Columns, 3µm Particle Size
Manufactured, bonded, tested in-house
Manufacturer:  Restek™ 9210311
 View more versions of this product
Catalog No. 06-717-043
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Pinnacle™ II Silica
No
50μL/min.
100mm
Spherical
2.5 to 8
80°C
L3
Pinnacle™ II Silica
1mm
3μm
Non basic compound analysis
Pinnacle™ II Silica
3μm
110Å
HPLC Column
HPLC