Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Poly Crimp Seal Caps (11mm)

Works as either a snap-on or crimp-top cap

Manufacturer:  Restek™ 24433

 View more versions of this product

Catalog No. 06-718-863


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Poly Crimp Seal
Poly
100 Pack
Poly
11mm
CONEG
Clear
Seal
PTFE/Butyl Rubber
2mL
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'