Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials
Manufacturer: Restek™ 24519
 View more versions of this product
Catalog No. 06-718-934
  Check Availability
Add to Cart
    Specifications
    Vial Volume (Metric) 250μL
    Material Polypropylene
    Type Vial Insert; Big mouth with top flange
    Capacity (Metric) 250μL
  • Description & Specifications
Related Products
Product Content Feedback