Learn More
Restek™ Porapak R Porous Polymers Packed GC Columns Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Restek™ 80470840901
 View more versions of this product
Catalog No. 06-714-650
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PerkinElmer Auto System 8300, 8400, 8700, Clarus 500 (C500)
1828.8mm
Porapak R Porous Polymer
80/100
3.175mm
250°C
Stainless Steel
Porapak R Porous Polymer
2.1mm