Restek™ Porous Polymers: HayeSep Series
Porous polymer adsorbent packing for GC columns.
Manufacturer: Restek™ 25036
 View more versions of this product
Catalog No. 06-712-611
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Quantity 24g
    Material Hayesep C
    Particle Size 80/100 Mesh
    Produce lower bleed and more reproducible characteristics.
  • Description & Specifications
Related Products
Product Content Feedback