Restek™ Porous Polymers: HayeSep Series
Porous polymer adsorbent packing for GC columns.
Manufacturer: Restek™ 25051
 View more versions of this product
Catalog No. 06-712-625
  Check Availability
Add to Cart
    Specifications
    Quantity 24g
    Material Hayesep R
    Particle Size 80/100 Mesh
    Produce lower bleed and more reproducible characteristics.
  • Description & Specifications
Related Products
Product Content Feedback