Restek™ Porous Polymers Packed Columns with SilcoSmooth™ Tubing Available on GSA/VA Contract for Federal Government customers only.
80/100 Mesh
Manufacturer: Restek™ 80434830
 View all options for this product
Catalog No. 06-714-567
  Check Availability
Add to Cart
    For Use With PE900-3920 Sigma 1,2,3
    Length (Metric) 2m
    Material SilcoSmooth™
    Technique GC
    Stationary Phase Chromosorb 102
    Packing Material Chromosorb 102
    Mesh Size 80/100
    Diameter (Metric) Inner 2mm
    Diameter (Metric) Outer 3.17mm
  • Description & Specifications
Related Products
Product Content Feedback