Restek™ Porous Polymers Packed Columns with SilcoSmooth™ Tubing Available on GSA/VA Contract for Federal Government customers only.
80/100 Mesh
Manufacturer: Restek™ 80435820
 View more versions of this product
Catalog No. 06-714-571
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With Varian 3700, Vista Series FID
    Length (Metric) 2m
    Material SilcoSmooth™
    Technique GC
    Stationary Phase Chromosorb 101
    Packing Material Chromosorb 101
    Mesh Size 80/100
    Diameter (Metric) Inner 2mm
    Diameter (Metric) Outer 3.17mm
  • Description & Specifications
Related Products
Product Content Feedback