Learn More
SGE PTFE Fluorocarbon Resin-Tipped, Gas-Tight Syringes from Restek™
Suitable for gases or liquids, for maximum inertness.
$95.50 - $710.50
Specifications
For Use With (Application) | Suitable for gases or liquids and high accuracy of dispensed volumes |
---|---|
Product Type | SGE PTFE Tip, Gas-Tight Syringes |
Quantity | Ea. |
Catalog Number | Mfr. No. | Volume (Metric) | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Model | Price | Quantity & Availability | ||||
06-712-416
|
Restek Corporation
24727 |
25 μL | 25F-GT |
Each for $124.50
|
|
||||
06-712-415
|
Restek Corporation
24726 |
10 μL | 10R-GT |
Each for $112.00
|
|
||||
06-712-417
|
Restek Corporation
24728 |
25 μL | 25R-GT |
Each for $112.00
|
|
||||
06-712-414
|
Restek Corporation
24725 |
10 μL | 10F-GT |
Each for $95.50
|
|
||||
06-712-418
|
Restek Corporation
24729 |
50 μL | 50F-GT |
Each for $124.50
|
|
||||
06-712-419
|
Restek Corporation
24730 |
50 μL | 50R-GT |
Each for $106.90
|
|
||||
06-712-422
|
Restek Corporation
24734 |
100 μL | 100F-GT |
Each for $146.30
|
|
||||
06-712-423
|
Restek Corporation
24735 |
100 μL | 100R-GT |
Each for $123.50
|
|
||||
06-712-426
|
Restek Corporation
24738 |
250 μL | 250F-GT |
Each for $146.30
|
|
||||
06-712-431
|
Restek Corporation
24743 |
500 μL | 500R-GT |
Each for $126.60
|
|
||||
06-712-430
|
Restek Corporation
24742 |
500 μL | 500F-GT |
Each for $99.60
|
|
||||
06-712-427
|
Restek Corporation
24739 |
250 μL | 250R-GT |
Each for $131.70
|
|
||||
06-712-441
|
Restek Corporation
24755 |
2.5 mL | 2.5MDF-LL-GT |
Each for $131.70
|
|
||||
06-712-436
|
Restek Corporation
24750 |
1 mL | 1MR-GT |
Each for $153.50
|
|
||||
06-712-443
|
Restek Corporation
24757 |
5 mL | 5MDR-LL-GT |
Each for $134.90
|
|
||||
06-712-438
|
Restek Corporation
24752 |
1 mL | 1MDF-LL-GT |
Each for $142.10
|
|
||||
06-712-446
|
Restek Corporation
24760 |
25 mL | 25MDR-LL-GT |
Each for $361.00
|
|
||||
06-712-445
|
Restek Corporation
24759 |
10 mL | 10MDR-LL-GT |
Each for $164.90
|
|
||||
06-712-447
|
Restek Corporation
24761 |
50 mL | 50MR-LL-GT |
Each for $432.50
|
|
||||
06-712-448
|
Restek Corporation
24762 |
100 mL | 100MR-LL-GT |
Each for $710.50
|
|
||||
Description
Suitable for gases or liquids, for maximum inertness.
- High accuracy of dispensed volumes
- Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
Suitable for gases or liquids and high accuracy of dispensed volumes | |
SGE PTFE Tip, Gas-Tight Syringes | |
Ea. |
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'