Learn More
Restek™ PTFE Fluorocarbon Resin Ferrules
One-piece design requires no back ferrule.
Manufacturer:  Restek™ 21124
 View more versions of this product
Catalog No. 06-718-430
  Check Availability
Add to cart
Specifications
Description
  • Completely inert
  • Upper temperature limit 250°C
Description & Specifications
Specifications
Ferrule
0.05cm
0.15cm
250°C
0.019 in.
0.0625 in.
482°F
PTFE Fluorocarbon Resin