Restek™ Q-sep™ QuEChERS dSPE Tubes for Extract Clean-Up

Fast, simple sample prep for multiresidue pesticide analysis.

Manufacturer: Restek™ 26123

 View more versions of this product

Catalog No. 06-729-300

  Check Availability
Add to Cart

    Specifications

    Product Type Q-sep™ QuEChERS dSPE micro-centrifuge tube
    Standard Components 150mg MgSO4, 50mg PSA, 50mg GCB
    Volume (Metric) 2mL
    For Use With (Application) Fast, simple sample prep for multi residue pesticide analysis
    Material 150mg MgSO4, 50mg PSA, 50mg GCB
    Method AOAC 2007.01
    Sample Volume 1mL
    Shake Time 2 min.
    Sorbent Ultra pure, PSA, GCB

    QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep products make QuEChERS procedures even simpler. All extraction salts, sorbents, and sample tubes are included. Multiple sorbents are used to extract different types of interferences.
    • Ready-to-use tubes, no glassware required
    • Preweighed, ultra-pure sorbents
    • Packaged in foil subpacks of 10 for enhanced protection and storage stability
    • Support original unbuffered, AOAC (2007.01), European (EN 15662), and mini-multiresidue QuEChERS methods

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit