Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rechargeable S-Traps

Individually activated for maximum reactivity

$223.00

Specifications

Length (English) 6.75 in.
Length (Metric) 17.2 cm
Material Brass
View More Specs

Products 1
Catalog Number Mfr. No. Connection Type Diameter (English) Outer Diameter (Metric) Outer Type Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Diameter (English) Outer Diameter (Metric) Outer Type Price Quantity & Availability  
06-711-821
View Documents
Restek™
22016
0.12 in. Fitting 5.62 in. 14.3 cm Hydrocarbon S Trap
Each for $223.00
Add to cart
 
Description

Description

Molecular Sieve S-Traps reduce water vapor to less than 1ppm. Use to increase column and oxygen trap lifetime, and reduce baseline noise for electrochemical detectors (ECD) and mass spectrometers. Available in 1/8 or 1/4 in. fittings.
Hydrocarbon S-Traps remove hydrocarbons and other contaminants from the gas stream. Reduce organics to 0.1ppm where input is 100ppm. 1/8 in. fittings only.

Order Info

Dimensions for both S-traps: 6 3/4L x 5 5/8 in. dia. (17.2 x 14.3cm).

Specifications

Specifications

6.75 in.
17.2 cm
Brass
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit