Learn More
Restek™ Rt-CW20M F and F Columns
Application-specific fused silica columns for flavor and fragrance compounds, specially tested.
Manufacturer:  Restek™ 12523
 View more versions of this product
Catalog No. 06-709-791
  Check Availability
Add to cart
Specifications
Description
  • Polar phase
  • Carbowax polyethylene glycol - not bonded
  • True nonbonded Carbowax 20M polarity
  • Mesh Size: 100/120
  • Temperature Range: 60° to 220/250°C
Description & Specifications
Specifications
Used for flavor and fragrance compound
0.25μm
0.25μm
Rt™-CW20M F and F
0.25mm
230°C
20 to 220°C
30m
Rt™-CW20M F and F