Learn More
Restek™ Rt™-Msieve 5A (Fused Silica PLOT) Columns Available on GSA/VA Contract for Federal Government customers only.
Efficient separation of Ar/O2 and other permanent gases, including CO
Manufacturer:  Restek™ 197236850
 View more versions of this product
Catalog No. 06-710-469
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
HP6850
50μm
30m
0.53mm