Learn More
Restek Rtx-1 Capillary Columns
Similar in polarity to DB-1 and HP-1
$552.00 - $5198.00
Specifications
Coating | Fused Silica |
---|---|
Packing Material | Rtx™-1 |
For Use With (Application) | Drugs of Abuse, Essential Oils, Gases Analysis, Hydrocarbons, Natural Gas Odorants Analysis, Oxygenates Analysis, PCB Congener Analysis, Pesticide Analysis, Semivolatiles Analysis, Simulated Distillation, Solvent Impurities Analysis, Sulfur Compounds Analysis |
Material | Stainless Steel |
Max. Temperature | 350°C |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
06-708-229
|
Restek
10108 |
0.25 mm | 0.10 µm df | 30 m | -60°C to +330/350°C |
Each for $606.00
|
|
06-610-614
|
Restek
10173 |
0.53 mm | 1.5 μm | 60 m | -60°C to 310°C to 330°C |
Each for $1,073.00
|
|
06-708-233
|
Restek
10109 |
0.32 mm | 0.1 µm | 30 m | -60°C to +330/350°C |
Each for $640.00
|
|
06-708-263
|
Restek
10123 |
0.25 mm | 0.25 µm | 30 m | -60°C to +330/350°C |
Each for $591.00
|
|
06-708-273
|
Restek
10124 |
0.32 mm | 0.25 µm df | 30 m | -60°C to +330/350°C |
Each for $623.00
|
|
06-708-361
|
Restek
10153 |
0.25 mm | 1 µm | 30 m | -60°C to +320/340°C |
Each for $587.00
|
|
06-708-366
|
Restek
10154 |
0.32 mm | 1 µm | 30 m | -60°C to +320/340°C |
Each for $623.00
|
|
06-708-420
|
Restek
10170 |
0.53 mm | 1.5 µm | 30 m | -60°C to +310/330°C |
Each for $708.00
|
|
06-708-441
|
Restek
10178 |
0.32 mm | 5 µm | 30 m | -60°C to +260/280°C |
Each for $622.00
|
|
06-708-446
|
Restek
10179 |
0.53 mm | 5 µm | 30 m | -60°C to +270/290°C |
Each for $708.00
|
|
06-708-479
|
Restek
10185 |
0.53 mm | 3 µm | 30 m | -60°C to +270/290°C |
Each for $701.00
|
|
06-610-588
|
Restek
10111 |
0.25 mm | 0.1 μm | 60 m | -60°C to +330/350°C |
Each for $974.00
|
|
06-610-591
|
Restek
10123600 |
0.25 mm | 0.25 μm | 30 m | -60°C to +330/350°C |
Pack of 6 for $2,755.00
|
|
06-610-593
|
Restek
10126 |
0.25 mm | 0.25 μm | 60 m | -60°C to +330/350°C |
Each for $959.00
|
|
06-610-595
|
Restek
10129 |
0.25 mm | 0.25 μm | 105 m | -60°C to +330/350°C |
Each for $1,106.00
|
|
06-610-599
|
Restek
10138 |
0.25 mm | 0.5 μm | 30 m | -60°C to +330/350°C |
Each for $552.00
|
|
06-610-601
|
Restek
10141 |
0.25 mm | 0.5 μm | 60 m | -60°C to +330/350°C |
Each for $974.00
|
|
06-610-604
|
Restek
10144 |
0.25 mm | 0.5 μm | 105 m | -60°C to +330/350°C |
Each for $1,106.00
|
|
06-610-589
|
Restek
10112 |
0.32 mm | 0.1 μm | 60 m | -60°C to +330/350°C |
Each for $1,039.00
|
|
06-610-602
|
Restek
10142 |
0.32 mm | 0.5 μm | 60 m | -60°C to +330/350°C |
Each for $1,039.00
|
|
06-610-608
|
Restek
10154600 |
0.32 mm | 1 μm | 30 m | -60°C to +320/340°C |
Pack of 6 for $2,933.00
|
|
06-708-376
|
Restek
10155 |
0.53 mm | 1 µm | 30 m | -60°C to +310/330°C |
Each for $706.00
|
|
06-610-610
|
Restek
10160 |
0.32 mm | 1 μm | 105 m | -60°C to +320/340°C |
Each for $1,261.00
|
|
06-610-612
|
Restek
10169 |
0.32 mm | 1.5 μm | 30 m | -60°C to +310/330°C |
Each for $588.00
|
|
06-610-613
|
Restek
10172 |
0.32 mm | 1.5 μm | 60 m | -60°C to +310/330°C |
Each for $1,039.00
|
|
06-610-615
|
Restek
10175 |
0.32 mm | 1.5 μm | 105 m | -60°C to +310/330°C |
Each for $1,261.00
|
|
06-610-621
|
Restek
10184 |
0.32 mm | 3 μm | 30 m | -60°C to +280/300°C |
Each for $588.00
|
|
06-610-623
|
Restek
10190 |
0.32 mm | 3 μm | 105 m | -60°C to +260/280°C |
Each for $1,261.00
|
|
06-610-627
|
Restek
10198 |
0.32 mm | 4 μm | 30 m | -60°C to +280/300°C |
Each for $617.00
|
|
06-610-618
|
Restek
10180600 |
0.32 mm | 5 μm | 60 m | -60°C to +260/280°C |
Pack of 6 for $5,198.00
|
|
06-610-587
|
Restek
10110 |
0.53 mm | 0.1 μm | 30 m | -60°C to +320/340°C |
Each for $659.00
|
|
06-610-592
|
Restek
10125 |
0.53 mm | 0.25 μm | 30 m | -60°C to +320/340°C |
Each for $659.00
|
|
06-610-594
|
Restek
10128 |
0.53 mm | 0.25 μm | 60 m | -60°C to +320/340°C |
Each for $1,073.00
|
|
06-610-600
|
Restek
10140 |
0.53 mm | 0.5 μm | 30 m | -60°C to +310/330°C |
Each for $659.00
|
|
06-610-603
|
Restek
10143 |
0.53 mm | 0.5 μm | 60 m | -60°C to +310/330°C |
Each for $1,073.00
|
|
06-610-609
|
Restek
10158 |
0.53 mm | 1 μm | 60 m | -60°C to +310/330°C |
Each for $1,089.00
|
|
06-610-622
|
Restek
10188 |
0.53 mm | 3 μm | 60 m | -60°C to +270/290°C |
Each for $1,089.00
|
|
06-610-617
|
Restek
10179600 |
0.53 mm | 5 μm | 30 m | -60°C to +270/290°C |
Pack of 6 for $3,288.00
|
|
06-610-625
|
Restek
10192 |
0.53 mm | 7 μm | 30 m | -60°C to +240/260°C |
Each for $659.00
|
|
06-610-626
|
Restek
10193 |
0.53 mm | 7 μm | 60 m | -60°C to +240/260°C |
Each for $1,104.00
|
|
Description
- Nonpolar; coated with crossbonded polydimethylsiloxane (100% dimethylpolysiloxane)
- Manufactured from high-quality fused silica tubing
- Surrounded by a rugged stainless-steel cage
- Excellent thermal stability for low bleed at high temperatures
Solvents, Petroleum products, Pharmaceuticals, Waxes, Fuel oils
Specifications
Fused Silica | |
Drugs of Abuse, Essential Oils, Gases Analysis, Hydrocarbons, Natural Gas Odorants Analysis, Oxygenates Analysis, PCB Congener Analysis, Pesticide Analysis, Semivolatiles Analysis, Simulated Distillation, Solvent Impurities Analysis, Sulfur Compounds Analysis | |
350°C | |
100% Dimethyl Polysiloxane |
Rtx™-1 | |
Stainless Steel | |
Non polar | |
G1, G2, G38 |