Learn More
Restek™ RTX™-1 Capillary Columns, 30m, 0.32mm ID and Under Available on GSA/VA Contract for Federal Government customers only.
Fused silica
Manufacturer:  Restek™ 101846850
 View more versions of this product
Catalog No. 06-708-478
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
G1, G2, G38
-60 to 280/300°C
30m
Crossbond™
Rtx™-1
Fused Silica
Not Applicable
350°C
3μm
3μm
Stainless Steel
100% Dimethyl Polysiloxane
Non polar
0.32mm