Learn More
Restek™ RTX™-1 Capillary Columns, 30m, 0.53mm ID or Larger Available on GSA/VA Contract for Federal Government customers only.
General-purpose columns for solvent impurities
Manufacturer:  Restek™ 101706850
 View more versions of this product
Catalog No. 06-708-425
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
G1, G2, G38
-60 to 310/330°C
30m
Crossbond™
Rtx™-1
Fused Silica
Cage
350°C
1.5μm
1.5μm
Stainless Steel
100% Dimethyl Polysiloxane
Non polar
0.53mm