Learn More
Restek™ RTX™-1 Capillary Columns, Under 30m, Particle Size 1.50µm or Less: I.D. 0.32mm Available on GSA/VA Contract for Federal Government customers only.
Fused silica
Manufacturer:  Restek™ 10151107
 View more versions of this product
Catalog No. 06-708-353
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
G1, G2, G38
-60 to 320/340°C
10m
Crossbond™
Rtx™-1
Fused Silica
Not Applicable
350°C
1μm
1μm
Stainless Steel
100% Dimethyl Polysiloxane
Non polar
0.32mm