Learn More
Restek™ RTX™-1701 Capillary Columns, Length 30m or Less: I.D. 0.32mm Available on GSA/VA Contract for Federal Government customers only.
General-purpose columns for alcohols, oxygenates, PCB congeners (e.g. Aroclor mixes), pesticides
Manufacturer:  Restek™ 12024106
 View more versions of this product
Catalog No. 06-709-544
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Alcohols, Oxygenates, PCB congeners, pesticides analysis
G46
-20 to 280°C
Cyano and Phenyl
Stainless Steel
14% Cyanopropylphenyl/86% Dimethyl Polysiloxane
Mid polar
0.32mm
350°C
0.25μm
0.25μm
25m
Crossbond™
Rtx™-1701
Fused Silica