Learn More
Restek™ Rtx™-20 Capillary Columns - 30m Length Available on GSA/VA Contract for Federal Government customers only.
General-purpose columns for volatile and flavor compounds.
Manufacturer:  Restek™ 10308
 View more versions of this product
Catalog No. 06-708-769
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatile compounds, flavor compounds, Alcoholic beverages analysis
G28, G32
-20 to 300/320°C
30m
Crossbond™
Rtx™-20
Fused Silica
350°C
0.1μm
0.1μm
Stainless Steel
20% Diphenyl/80% Dimethyl Polysiloxane
Low to mid polar
0.25mm