Learn More
Restek™ Rtx-20 GC Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Rtx-20 polymer is synthesized to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight monomodal distribution and extremely low bleed.
Manufacturer:  Restek™ 10320
 View more versions of this product
Catalog No. 06-708-777
  Check Availability
Add to cart
Specifications
Description
  • General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
  • Equivalent to USP G28, G32 phases.
Description & Specifications
Specifications
Volatile compounds, flavor compounds, Alcoholic beverages analysis
350°C
0.25μm
0.25μm
Stainless Steel
20% Diphenyl/80% Dimethyl Polysiloxane
Low to mid polar
0.25mm
Capillary Column
G28, G32
-20 to 300/320°C
15m
Crossbond™
Rtx™-20
Fused Silica
1 Ea.