Learn More
Restek™ Rtx™-200 Capillary Columns - 15m Length Available on GSA/VA Contract for Federal Government customers only.
General-purpose, fused silica columns for solvents, Freon fluorocarbons, alcohols, ketones, silanes, glycols, and drugs of abuse. Restek Rtx-200 Capillary Columns have excellent confirmation with Rtx-5 columns for phenols, nitrosamines, organochlorine pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides.
Manufacturer:  Restek™ 15037
 View more versions of this product
Catalog No. 06-710-131
  Check Availability
Add to cart
Specifications
Description
Rtx-200 columns have accomplished many difficult separations not possible on any other bonded stationary phase. The Rtx-200 column offers exceptional thermal stability, low bleed, and superior inertness, even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs.
  • Inert, midpolarity
  • Crossbond trifluoropropylmethyl polysiloxane stationary phase has a unique selectivity that changes elution orders and resolves compounds that phenyl, cyano, or Carbowax phases cannot
  • Equivalent to USP G6 phase
Order Info
Custom lengths and film thicknesses available.
Description & Specifications
Specifications
Solvents, Freon™ Fluorocarbons, Alcohols, Ketones, Silanes, Glycols analysis
G6
-20° to +300/320°C
15m
Crossbond™
Rtx™-200
Fused Silica
350°C
0.5μm
0.5μm
Stainless Steel
Trifluoropropylmethyl Polysiloxane
Mid polar
0.53mm