Learn More
Restek™ Rtx™-200 Capillary Columns - 60m Length Available on GSA/VA Contract for Federal Government customers only.
Excellent confirmation to the Rtx-5 column for phenols, pesticides and herbicides
Manufacturer:  Restek™ 150726850
 View more versions of this product
Catalog No. 06-710-177
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvents, Freon™ Fluorocarbons, Alcohols, Ketones, Silanes, Glycols analysis
G6
-20 to 280/300°C
60m
Crossbond™
Rtx™-200
Fused Silica
Cage
350°C
1.5μm
1.5μm
Stainless Steel
Trifluoropropylmethyl Polysiloxane
Mid polar
0.32mm