Learn More
Restek™ Rtx™-200 Capillary Columns: Length 30m, with Guard or Cage Available on GSA/VA Contract for Federal Government customers only.
Excellent confirmation to the Rtx-5 column for phenols, pesticides and herbicides
Manufacturer:  Restek™ 150396850
 View more versions of this product
Catalog No. 06-710-135
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Solvents, Freon™ Fluorocarbons, Alcohols, Ketones, Silanes, Glycols analysis
G6
-20° to +310/330°C
30m
Crossbond™
Rtx™-200
Fused Silica
Cage
350°C
0.5μm
0.5μm
Stainless Steel
Trifluoropropylmethyl Polysiloxane
Mid polar
0.32mm