Learn More
Restek™ RTX™-225 Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Similar to DB-225 and HP-225 columns
Manufacturer:  Restek™ 140396850
 View more versions of this product
Catalog No. 06-710-053
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
FAMEs, Carbohydrates, Sterols, flavor compounds analysis
G7, G19
40 to 220/240°C
30m
Crossbond™
Rtx™-225
Fused Silica
Cage
350°C
0.5μm
0.5μm
Stainless Steel
50% Cyanopropylmethyl/50% Phenylmethyl Polysiloxane
Polar
0.32mm