Learn More
Restek™ RTX™-5 Amine Capillary Columns: With Guard or Cage Available on GSA/VA Contract for Federal Government customers only.
Tubing surface is chemically altered to eliminate tailing of basic compounds
Manufacturer:  Restek™ 123536850
 View more versions of this product
Catalog No. 06-709-724
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Amines, Alkylamines, Diamines, Triamines, Ethanolamines, Nitrogen containing Heterocyclics analysis
1μm
1μm
Stainless Steel
5% Diphenyl/95% Dimethyl Polysiloxane
Low polar
0.25mm
350°C
-60 to 300/315°C
30m
Crossbond™
Rtx™-5 Amine
Fused Silica
Cage